Learn More
Thermo Scientific Chemicals Calpain Inhibitor II, 95+%
Beschreibung
ALLM (Calpain Inhibitor) is a cell-permeable, peptide aldehyde inhibitor of calpain I and calpain II. ALLM acts as a very potent inhibitor of cathepsin L and the strongest inhibitor of cathepsin B amongst the peptide aldehydes. This compound inhibits the processing of malaria aspartic hemoglobinases plasmepsins I and II in vitro. ALLM also inhibits other neutral cysteine proteases, and activation-induced programmed cell death. It restores defective immune responses in HIV+ donors. ALLM blocks nitric oxide production by activated macrophages by interfering with transcription of the inducible nitric oxide synthase gene. ALLM also acts as a weak inhibitor of proteasome.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Quantity | 25 mg |
| Description | White |
| Content And Storage | -30°C to -10°C |
| For Use With (Application) | An inhibitor of calpains I and II |
| Formula Weight | 401.60g/mol |
| Solubility | Soluble in ethanol at 20mg/ml |
| Product Type | Signal Transduction Reagent |
| Form | Powder |
| Assay | ≥95% |
RUO – Research Use Only
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.