missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Calpain Inhibitor XII

Product Code. 15496559
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15496559 1 mg 1mg
15486559 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496559 Supplier Thermo Scientific Chemicals Supplier No. J64751.M^
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

A cell-permeable dipeptidyl --ketoamide compound that acts as a potent, selective, reversible, and active site inhibitor of calpain-1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Description Pale yellow
Content And Storage 2°C to 8°C
For Use With (Application) A cell-permeable dipeptidyl →-ketoamide compound that acts as a potent, selective, reversible, and active site inhibitor of calpain-1
Molecular Formula C26H34N4O5
Formula Weight 482.60
Synonym Z-L-Nva-CONH-CH2-2-Py
Form Solid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.