missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Calpeptin, ≥98%

Product Code. 15443249
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15443249 10 mg 10mg
15453249 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15443249 Supplier Thermo Scientific Chemicals Supplier No. J60481.MA
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibitor of the calcium-dependent protease calpain and cathepsin L

Calpeptin is used to to inhibit the activities of cathepsin L and calpain-1. Calpeptin blocked the expression of aspartyl protease cathepsin D, amyloid precursor protein, and calparin. It acts as an inhibitor of the Ca2+-dependent protease, calpain. Prevents collagen- and thrombin-induced platelet aggregation, probably by blocking calpain induced phospholipase C and thromboxane synthase activation. Potent cathepsin L inhibitor. It is shown to preferentially inhibit a subset of protein-tyrosine phosphatases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117591-20-5
Molecular Formula C20H30N2O4
Synonym N-Benzyloxycarbonyl-L-leucyl-norleucinal; Z-Leu-norleucinal

Specifications

Quantity 10 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) Inhibitor of the calcium-dependent protease calpain and cathepsin L
Formula Weight 362.50g/mol
Solubility Soluble in DMSO at 15mg/ml
Product Type Signal Transduction Reagent
Form Powder
Assay ≥98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.