missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Cam Lock

Product Code. 16309225 Shop All MP Biomedicals Products
Change view
Click to view available options
For Use With (Equipment):
FastPrep-24™ 5G Instruments
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16309225 FastPrep-24™ 5G Instruments Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16309225 Supplier MP Biomedicals™ Supplier No. 116005588
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cam Lock is a locking system for FastPrep-24™ 5G Instruments.

Modeled after the bike tire quick release, the familiar mechanism allows:
  • Rapid sample exchange
  • Strong grip
  • Uniform clamping force
  • Ease of use

Specifications

For Use With (Equipment) FastPrep-24™ 5G Instruments
Packaging Quantity 1 Pk.

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.