missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Camostat mesylate

Product Code. 18782494
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18782494 50 mg 50mg
18712614 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18782494 Supplier Tocris Bioscience™ Supplier No. 3193/50
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Orally active protease inhibitor

Camostat mesylate is an orally active protease inhibitor. Inhibits trypsin and various inflammatory proteases including plasmin, kallikrein and thrombin. Suppresses pancreatitis-induced pain in rats following oral administration. Also inhibits transmembrane serine protease TMPRSS2 and partially blocks entry of SARS-Cov-2 (COVID-19) into lung cells in vitro.

Tocris products are for biomedical research use only. They are not intended for human or veterinary use.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59721-29-8
Molecular Formula C21H26N4O8S

Specifications

Quantity 50 mg
Formula Weight 494.52
Purity >98%
Molecular Weight (g/mol) 494.52
Chemical Name or Material Camostat mesylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.