missing translation for 'onlineSavingsMsg'
Learn More

Camostat methanesulfate, 98%, Thermo Scientific™

An orally active synthetic serine protease inhibitor

Brand:  Thermo Scientific Alfa Aesar J62362.MC

Quantity 100mg

 View more versions of this product

Product Code. 15434759

  • 284.00 € / 100mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as a protease and Trypsin inhibitor. Furthermore it was found that this water induction and bicarbonate secretion increase was achieved via direct regulation of ductular cells of the dog pancreas. Other studies have shown Camostat mesylate to suppress spinal Fos expression in the rat, which is a marker for neuronal activation. In addition it was shown that this compound may prevent or depress pancreatitis-induced referred allodynia/hyperalgesia in rat models. Camostat mesylate has also demonstrated the ability to prevent Trypsin attack of microspheres, which causes the degradation product N-benzoyl-dl-arginine. Camostat mesylate is an inhibitor of KLK, plasminogen, Prothrombin and Trypsin.

Solubility
Soluble in water (100 mM), DMSO (100 mM), and ethanol (<1mg/mL).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

59721-29-8
494.52
FSEKIHNIDBATFG-UHFFFAOYSA-N
5284360
CS(O)(=O)=O.CN(C)C(=O)COC(=O)CC1=CC=C(OC(=O)C2=CC=C(C=C2)N=C(N)N)C=C1
C21H26N4O8S
MFCD00941410
camostat mesylate, camostat mesilate, camostat methanesulfonate, camostat monomethanesulfonate, foipan, unii-451m50a1eq, dsstox_cid_238, dsstox_rid_75452, dsstox_gsid_20238
4-{2-[(dimethylcarbamoyl)methoxy]-2-oxoethyl}phenyl 4-[(diaminomethylidene)amino]benzoate; methanesulfonic acid

Specifications

59721-29-8
MFCD00941410
14,1728
Soluble in water (100 mM),DMSO (100 mM),and ethanol (<1mg/ml).
CS(O)(=O)=O.CN(C)C(=O)COC(=O)CC1=CC=C(OC(=O)C2=CC=C(C=C2)N=C(N)N)C=C1
494.52
494.52
Camostat methanesulfate
C21H26N4O8S
100mg
camostat mesylate, camostat mesilate, camostat methanesulfonate, camostat monomethanesulfonate, foipan, unii-451m50a1eq, dsstox_cid_238, dsstox_rid_75452, dsstox_gsid_20238
FSEKIHNIDBATFG-UHFFFAOYSA-N
4-{2-[(dimethylcarbamoyl)methoxy]-2-oxoethyl}phenyl 4-[(diaminomethylidene)amino]benzoate; methanesulfonic acid
5284360
98%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.