missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ cAMP Parameter Assay Kit
Click to view available options
Quantity:
1 Kit
1 Pack
Sufficient For:
1 x 96-well plate
6 x 96-well plates
Unit Size:
1 set
Description
The cAMP Parameter Assay Kit quantifies multi-species cAMP.
Specifications
Specifications
| Assay Range | 3.75 to 240 pmol/mL |
| Assay Sensitivity | 8.57 pmol/mL |
| Conjugate | Biotin |
| Cross Reactivity | Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules. |
| Product Type | ELISA Kit |
| Sample Type | Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Saliva, Serum, Urine |
| Specificity | Cyclic AMP Levels in Various Samples |
| Sufficient For | 1 x 96-well plate |
| Target Species | Multi-species |
| For Use With (Application) | ELISA |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction