missing translation for 'onlineSavingsMsg'
Learn More

Campesterol-d7 (Mixture of Diastereomers), TRC

Product Code. 30335396
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30335396 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30335396 Supplier Toronto Research Chemicals Supplier No. TRCC15536410MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Campesterol-d7 (Mixture of Diastereomers)
CAS 2483832-11-5
Molecular Formula C28 D7 H41 O
Quantity 10 mg
Synonym Ergost-5-en-25,26,26,26,27,27,27-d7-3-ol, (3β,24ξ)- (ACI), (3β,24ξ)-Ergost-5-en-25,26,26,26,27,27,27-d7-3-ol (ACI), (24ξ)-24-Methylcholest-5-en-3β-ol-25,26,26,26,27,27,27-d7, (3β,24ξ)-Ergost-5-en-3-ol-25,26,26,26,27,27,27-D7, 5-Cholesten-24(RS)-methyl-3β-ol-25,26,26,26,27,27,27-d7
InChI Formula InChI=1S/C28H48O/c1-18(2)19(3)7-8-20(4)24-11-12-25-23-10-9-21-17-22(29)13-15-27(21,5)26(23)14-16-28(24,25)6/h9,18-20,22-26,29H,7-8,10-17H2,1-6H3/t19?,20-,22+,23+,24-,25+,26+,27+,28-/m1/s1/i1D3,2D3,18D
SMILES [2H]C([2H])([2H])C([2H])(C(C)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C([2H])([2H])[2H]
IUPAC Name (3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6,7,7,7-tetradeuterio-5-methyl-6-(trideuteriomethyl)heptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
Molecular Weight (g/mol) 407.7233
Formula Weight 407.4145
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.