missing translation for 'onlineSavingsMsg'
Learn More

(-)-Camphanic acid chloride, 98%, Thermo Scientific Chemicals

Catalog Number 10398580
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
39637-74-6
C10H13ClO3
216.66
MFCD00135626
PAXWODJTHKJQDZ-QVDQXJPCSA-N
1s-4,7,7-trimethyl-3-oxo-2-oxabicyclo 2.2.1 heptane-1-carbonyl chloride, --1s,4r-camphanoyl chloride, 1s---camphanic acid chloride
12364938
(4S)-1,7,7-trimethyl-2-oxo-3-oxabicyclo[2.2.1]heptane-4-carbonyl chloride
CC1(C2(CCC1(OC2=O)C(=O)Cl)C)C
This item is not returnable. View return policy
39637-74-6
C10H13ClO3
216.66
MFCD00135626
PAXWODJTHKJQDZ-QVDQXJPCSA-N
1s-4,7,7-trimethyl-3-oxo-2-oxabicyclo 2.2.1 heptane-1-carbonyl chloride, --1s,4r-camphanoyl chloride, 1s---camphanic acid chloride
12364938
(4S)-1,7,7-trimethyl-2-oxo-3-oxabicyclo[2.2.1]heptane-4-carbonyl chloride
CC1(C2(CCC1(OC2=O)C(=O)Cl)C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 67.0°C to 71.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 1 g
Specific Rotation − 18.00
Solubility Information Solubility in water: decomposes
Formula Weight 216.66
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (-)-Camphanic acid chloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • (-)-Camphanic acid chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'