missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Camptothecin

Product Code. 16471806
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471806 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16471806 Supplier MedChemExpress Supplier No. HY16560
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Camptothecin (CPT), a kind of alkaloid, is a DNA topoisomerase I (Topo I) inhibitor with an IC50 of 679 nM. Camptothecin (CPT) exhibits powerful antineoplastic activity against colorectal, breast, lung and ovarian cancers, modulates hypoxia-inducible factor-1α (HIF-1α) activity by changing microRNAs (miRNA) expression patterns in human cancer cells.

Specifications

Content And Storage 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Molecular Weight (g/mol) 348.35 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 3-4-7689
Product Type Natural Products
Purity 96.17%
Solubility DMSO : 6.25 mg/mL (17.94 mM∣Need ultrasonic)
Synonym Campathecin∣ (S)-(+)-Camptothecin∣ CPT
Target Topoisomerase∣ADC Cytotoxin∣MicroRNA∣Influenza Virus∣Apoptosis∣Fungal∣Antibiotic
Quantity 500 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.