missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Can-Bus Cable Mastercycler™ EP 50cm

Product Code. 10219893 Shop All Eppendorf Products
Change view
Click to view available options
Product Type:
Can-bus Cable Mastercycler EP 50 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
10219893 Can-bus Cable Mastercycler EP 50 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10219893 Supplier Eppendorf™ Supplier No. 5341612006
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.