Learn More
Cadence Science™ Cancer Implant Needle
Product Code. 17129548
Change view
Click to view available options
Thickness:
11 ga
Unit Size:
Pack of 12
Product Code. Thickness Unit Size
17129548 11 ga Pack of 12
1 options
Product Code. 17129548 Supplier Cadence Science™ Supplier No. 7928
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Needle provides simple procedure to obtain and implant tumorous tissue cells beneath skin of lab animals for observation while maintaining a sterile field
  • Two piece needle set
  • Consists of sharp pointed stainless-steel outer cannula and stainless steel blunt obturator which protrudes 3.2mm beyond the needle point
Specifications
For Use With (Application) Obtain and implant tumorous tissue cells beneath the skin of lab animals
Type Cancer Implant Needle
Material Stainless Steel
Size Standard
Thickness 11 ga
Length (English) 3.25 in.
Length (Metric) 8.25 cm