missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Cannabigerol

Product Code. 18045302
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18045302

Brand: Tocris Bioscience™ 3021/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cannabinoid receptor partial agonist

Cannabigerol is a partial agonist of CB2 and CB1 receptors (Ki values are 337 and 440 nM respectively). Also exhibits α2-adrenoceptor agonist activity (EC50 = 0.2 nM in mouse brain membranes).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[(2E)-3,7-Dimethyl-2,6-octadienyl)-5-pentyl-1,3-benzenediol
CAS 25654-31-3
Quantity 10 mg
Target Non-selective Cannabinoid Receptor Agonists
Molecular Formula C21H32O2
Purity 95%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.