missing translation for 'onlineSavingsMsg'
Learn More

Poulten Selfe™ Cannon-Fenske Opaque Viscometer

Product Code. 13101093
Change view
Click to view available options
Product Type:
Cannon-Fenske Viscometer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
13101093 Cannon-Fenske Viscometer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13101093 Supplier Poulten Selfe™ Supplier No. 1641/07
  • ONLINE EXCLUSIVE PRICE
    208.00 €
    191.76 € / Pack of 1
    Save 16.24 €
    7% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Order Info

SampleVolumeMetric

Specifications

Product Type Cannon-Fenske Viscometer
Viscosity Range 50 to 200 cSt
For Use With (Application) Opaque Liquids
Size 300
Volume (Metric) 12 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.