Learn More
Paragon Scientific™ Cannon-Fenske Opaque Viscometer for Transparent and Opaque Liquids
Product Code. 15979959
Change view
Click to view available options
Viscosity Range:
0.4 to 2 cSt
0.8 to 4 cSt
1.6 to 8 cSt
10,000 to 50,000 cSt
100 to 500 cSt
1600 to 8000 cSt
20 to 100 cSt
20,000 to 100,000 cSt
240 to 1200 cSt
3 to 15 cSt
4000 to 20,000 cSt
50 to 100 cSt
500 to 2500 cSt
7 to 35 cSt
Unit Size:
Each
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Viscosity Range unitSize
15979959 0.4 to 2 cSt Each
15999959 0.8 to 4 cSt Each
15919969 1.6 to 8 cSt Each
15939969 3 to 15 cSt Each
15959969 7 to 35 cSt Each
15979969 20 to 100 cSt Each
15999969 50 to 100 cSt Each
15919979 100 to 500 cSt Each
15939979 240 to 1200 cSt Each
15959979 500 to 2500 cSt Each
15979979 1600 to 8000 cSt Each
15999979 4000 to 20,000 cSt Each
15919989 10,000 to 50,000 cSt Each
15939989 20,000 to 100,000 cSt Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 15979959 Supplier Paragon Scientific™ Supplier No. CFO01NC
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For the determination of kinematic viscosity of transparent and opaque Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
  • High Quality glass capillary viscometer
  • Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifications
Viscosity Range 0.4 to 2 cSt
Calibrated No
Approximate Constant 0.002
ASTM Numbers D 445, D 446
Certifications/Compliance ASTM D445, ISO 3104 and IP 71 Section 1
Size 25
4 week lead-time