missing translation for 'onlineSavingsMsg'
Learn More

Gravis Cap

Product Code. 11713863
Click to view available options
For Use With (Equipment):
25mm Thread Size
28mm Thread Size
35mm Thread Size
45mm Thread Size
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 11713863

Brand: Gravis P09025S100

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Specifications

No. per Pack 100Pack
Quantity 100Pack
Product Type Cap
For Use With (Equipment) 25mm Thread Size
Closure Size 25 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.