missing translation for 'onlineSavingsMsg'
Learn More

Capecitabine, MedChemExpress

Product Code. 16402039
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16402039 1 g 1g
16460758 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16402039 Supplier Medchem Express Supplier No. HYB00161g
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Capecitabine is an oral prodrug that is converted to its active metabolite, 5-FU, by thymidine phosphorylase.

Capecitabine is an oral prodrug that is converted to its active metabolite, 5-FU, by thymidine phosphorylase.

Specifications

Chemical Name or Material Capecitabine
CAS 154361-50-9
Color White
Purity Grade Notes Research
Molecular Formula C15H22FN3O6
Quantity 1 g
Solubility Information DMSO : 250 mg/mL (695.70 mM; Need ultrasonic) ∣H2O : ≥ 33.33 mg/mL (92.75 mM)
SMILES C[C@@H]1[C@H]([C@H]([C@H](N2C(N=C(C(F)=C2)NC(OCCCCC)=O)=O)O1)O)O
Molecular Weight (g/mol) 359.35
Formula Weight 359.35
Percent Purity 98.23%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.