missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Capillary Column End Caps

Product Code. 11827210 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
GC Capillary Columns
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11827210 GC Capillary Columns Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11827210 Supplier Thermo Scientific™ Supplier No. 260EC111
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
Estimated Shipment: 11-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eliminate column contamination with these universal, reusable, color-coded end caps.

Eliminate column contamination with Thermo Scientific™ Capillary Column End Caps. These universal and reusable fittings are color-coded to ensure that the column is reinstalled the same way it came out.

  • Eliminate column contamination caused by leaving a column unsealed or sealed with a septum
  • Color-coordinated fittings ensure that the column is reinstalled the same way it came out
  • Reusable
TRUSTED_SUSTAINABILITY

Specifications

Type End Caps
Description End Caps; For Capillary Columns
For Use With (Equipment) GC Capillary Columns

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.