Learn More
Marienfeld Superior™ Capillary Tubes for Microhaematocrits
Product Code. 13522140
Change view
Click to view available options
Capacity (Metric):
60 μL
75 μL
Diameter (Metric) Inner:
0.9 to 1.0 mm
1.1-1.2 mm
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Inner Unit Size
13522140 75 μL 1.1-1.2 mm Pack of 1000
13552140 60 μL 0.9 to 1.0 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13522140 Supplier Marienfeld Superior™ Supplier No. 2901000
  • ONLINE EXCLUSIVE PRICE
    31.64 €
    27.94 € / Pack of 1000
    Save 3.70 €
    11% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Use Marienfeld Superior™ Capillary tubes for microhaematocrits for the centrifugation of blood samples
  • Made of high grade quality glass
  • In compliance with DIN ISO 12 772
  • Available with and without heparinization
  • Fire polishing at the color coded end ensures that centrifuges are not damaged
  • For single use
  • 100 pieces in a dispenser vial, 10 vials in a box
  • For in-vitro diagnostic (IVD) applications according to IVD directive 98/79/EC, with CE-mark, recommended best-before-date and batch number for comprehensive information and traceability
Specifications
Product Type Capillary Tube
Additive Sodium Heparin
Material Glass
Capacity (Metric) 75 μL
Diameter (Metric) Inner 1.1-1.2 mm
Quantity 10x100
For Use With (Application) Centrifugation of blood samples
Length (Metric) 75 mm
Certifications/Compliance CE-marked (IVD)