Learn More
Thermo Scientific™ Capit-All Flex Cap Driver Cassettes
Product Code. 31872965 Shop All Thermo Scientific Products
Change view
Click to view available options
Compatible Storage Tubes:
Azenta 96 Format External Thread
Azenta 96 Format Internal Thread
Matrix and Nunc tubes in 96 or 48 formats
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Compatible Storage Tubes Unit Size
31872965 Azenta 96 Format External Thread Each
31872964 Azenta 96 Format Internal Thread Each
30274494 Matrix and Nunc tubes in 96 or 48 formats Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31872965 Supplier Thermo Scientific™ Supplier No. 4130FDXE
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Thermo Scientific Capit-All Flex Cap Driver Cassettes are compatible with both Matrix and Nunc tubes in 96 or 48 formats, and Azenta FluidX tubes in 96 format. It is recommended to be replaced after 10,000 decapping cycles.
  • Cap driver cassette compatible with Thermo Scientific Capit-All Flex Automated Decapper (P/N 4120-FLEX)
  • Tube compatibility: Matrix and Nunc cryotubes in 96 and 48 formats; Azenta FluidX tubes with internal threads in 96 format
  • Replace after 10,000 cycles
TRUSTED_SUSTAINABILITY
Specifications
Compatible Storage Tubes Azenta 96 Format External Thread
Decapping Format 96-Channel
Type Decapping Head