Description
The Thermo Scientific Capit-All Flex Cap Driver Cassettes are compatible with both Matrix and Nunc tubes in 96 or 48 formats, and Azenta FluidX tubes in 96 format. It is recommended to be replaced after 10,000 decapping cycles.
- Cap driver cassette compatible with Thermo Scientific Capit-All Flex Automated Decapper (P/N 4120-FLEX)
- Tube compatibility: Matrix and Nunc cryotubes in 96 and 48 formats; Azenta FluidX tubes with internal threads in 96 format
- Replace after 10,000 cycles
Specifications
Specifications
| Compatible Storage Tubes | Matrix and Nunc tubes in 96 or 48 formats, and Azenta FluidX tubes in 96 format |
| Decapping Format | 96-Channel |
| Operation | Touchscreen User Interface |
| Type | Decapping Head |