missing translation for 'onlineSavingsMsg'
Learn More

Caproic acid, ∽98%, MP Biomedicals™

Product Code. 11427851 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL

Product Code. 11427851

Brand: MP Biomedicals 0215055380

Please to purchase this item. Need a web account? Register with us today!

Specifications

CAS 142-62-1
Color Colorless
Density 0.927 (water=1)
Assay Percent Range ∽98%
Molecular Formula C6H12O2
Quantity 100 mL
Synonym caproic acid, capronic acid, n-hexanoic acid, n-caproic acid, butylacetic acid, hexoic acid, pentylformic acid, 1-hexanoic acid, n-hexylic acid, n-hexoic acid
InChI Key FUZZWVXGSFPDMH-UHFFFAOYSA-N
SMILES CCCCCC(=O)O
IUPAC Name hexanoic acid
Molecular Weight (g/mol) 116.16
PubChem CID 8892
ChEBI CHEBI:30776
Formula Weight 116.18
Physical Form Liquid
Chemical Name or Material Caproic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.