missing translation for 'onlineSavingsMsg'
Learn More

epsilon-Caprolactone monomer, 99%

Product Code. 10173713 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
250g
Product Code. Quantity Packaging unitSize
10173713 2.5 kg Glass bottle 2.5kg
10092080 250 g Glass bottle 250g
10742601 1 kg Glass bottle 1kg
10388539 10 kg Plastic drum 10kg
4 options
This item is not returnable. View return policy

Product Code. 10173713

Brand: Thermo Scientific Acros 173440025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 502-44-3 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 502-44-3
MDL Number MFCD00003267
InChI Key PAPBSGBWRJIAAV-UHFFFAOYSA-N
Synonym 6-hexanolactone, epsilon-caprolactone, 2-oxepanone, caprolactone, hexan-6-olide, 6-hexanolide, hexano-6-lactone, 1,6-hexanolide, e-caprolactone, 1-oxa-2-oxocycloheptane
PubChem CID 10401
ChEBI CHEBI:17915
IUPAC Name oxepan-2-one
SMILES C1CCC(=O)OCC1

Specifications

Color Colorless
Density 1.0300g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C6H10O2
Refractive Index 1.4620 to 1.4640 (20°C, 589nm)
Quantity 2.5 kg
Molecular Weight (g/mol) 114.144
Formula Weight 114.14
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material ε-Caprolactone monomer
Show More Show Less
compliance-icons
Product Identifier
  • e-Caprolactone monomer
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.