Learn More
Thermo Scientific™ Caps for Nunclon™ Delta Flasks
BIOPHARMA
Product Code. 10654492
Change view
Click to view available options
For Use With (Equipment):
175 cm² EasY Flask
175/185/500 cm² Nunclon Delta Flask
25 cm² EasY Flask
25 cm² Nunclon Delta Flask
75 cm² EasY Flask
80 cm² Nunclon Delta Flask
Unit Size:
Pack of 100
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10654492 175 cm² EasY Flask Pack of 100
10357351 25 cm² EasY Flask Pack of 100
10574751 75 cm² EasY Flask Pack of 100
10154271 25 cm² Nunclon Delta Flask Pack of 100
10203981 80 cm² Nunclon Delta Flask Pack of 100
10090611 175/185/500 cm² Nunclon Delta Flask Pack of 100
10584751 75 cm² EasY Flask Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10654492 Supplier Thermo Scientific™ Supplier No. 133001
  • ONLINE EXCLUSIVE PRICE
    110.00 €
    70.26 € / Pack of 100
    Save 39.74 €
    36% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Protect your valuable cell culture samples. These caps for Thermo Scientific™ Nunclon™ Delta Flasks are offered in two styles—vent/close, filter—to meet application needs.
Protect your valuable cell culture samples. These caps for Thermo Scientific™ Nunclon™ Delta Flasks are offered in two styles—vent/close (solid) and filter—to meet your application needs.
Vent/close Caps
  • EasY Flask vent/close caps feature a "Y" mark that allows visual verification
  • Vent position easily distinguised even what flasks are stacked in incubators
Filter Caps
  • Filter membrane provides effective barrier against microorganisms
  • Well-defined, hydrophobic filter membrane offers continuous venting with consistent gas exchange and no lot-to-lot variation
  • Chemically robust membrane meets USP class VI standards and is produced without animal-derived substances
Certifications:
  • Sterilized by gamma irradiation and certified to be nonpyrogenic
Ordering Information:
  • Case of 100
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Blue
Description Sterile, HDPE closure
Material HDPE
For Use With (Equipment) 175 cm² EasY Flask
No. per Case 100
No. per Pack 1
Product Type Vent/close cap
Sterility Sterile
Closure Type Solid
Culture Area 175 cm2, 225 cm2
Surface Treatment Nunclon Delta
Show More Show Less
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.