missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals CAPSO, 99%

Product Code. 11965421 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
1kg
25g
This item is not returnable. View return policy

Product Code. 11965421

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 73463-39-5 | C9H19NO4S | 237.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73463-39-5
Molecular Formula C9H19NO4S
Molecular Weight (g/mol) 237.3
InChI Key INEWUCPYEUEQTN-UHFFFAOYSA-N
Synonym capso, 3-cyclohexylamino-2-hydroxy-1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxypropane-1-sulfonic acid, capso free acid, 1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxy, capso compound, 3-cyclohexylamino-2-hydroxy-1-propanesuhicic acid, 3-cyclohexylamino-2-hydroxy-1-propanesulphonic acid, 3-cyclohexylamino-2-hydroxypropanesulfonic acid
PubChem CID 2733480
IUPAC Name 3-(cyclohexylamino)-2-hydroxypropane-1-sulfonic acid
SMILES C1CCC(CC1)NCC(CS(=O)(=O)O)O

Specifications

Absorbance (0.1M aq. soln.), 0.03 max. at 280nm, (0.1 M aq. soln.), (0.1 M aq. soln.) at 0nm
Melting Point 263.0°C to 271.0°C
Color White
pH 5 to 6.5 (1% aq. soln. at 25°C)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 1 kg
Solubility Information (0.1M in water) Clear colorless solution
Formula Weight 237.3
Percent Purity 99%
Physical Form Powder
Chemical Name or Material CAPSO
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.