missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals CAPSO sodium salt, 98%

Product Code. 11985291 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
Product Code. Quantity Packaging unitSize
11985291 25 g Glass bottle 25g
11995291 100 g Glass bottle 100g
2 options
This item is not returnable. View return policy

Product Code. 11985291

Brand: Thermo Scientific Chemicals 444130250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102601-34-3
Molecular Formula C9H12NO4S
Molecular Weight (g/mol) 230.26
MDL Number MFCD00070063
InChI Key CLEVULOKVPBHCU-VIFPVBQESA-M
Synonym capso sodium salt, sodium 3-cyclohexylamino-2-hydroxypropane-1-sulfonate, 3-cyclohexylamino-2-hydroxypropanesulfonic acid sodium salt, capso sodium, capso, 0.2m buffer solution, ph 8.5, capso, 0.2m buffer solution, ph 9.0, capso, 0.2m buffer solution, ph 9.5, sodium 3-cyclohexylamino-2-hydroxypropanesulfonate, sodium 3-cyclohexylamino-2-hydroxy-propane-1-sulfonate, potassium 3-cyclohexylamino-2-hydroxypropane-1-sulfonate
PubChem CID 25000353
IUPAC Name sodium;3-(cyclohexylamino)-2-hydroxypropane-1-sulfonate
SMILES O[C@@H](CNC1=CC=CC=C1)CS([O-])(=O)=O

Specifications

Absorbance 0.050 max. (280nm, 0.1M H2O), 0.050 max. (260nm, 0.1M H20), 0.050 max. (0.1M H2O) at 280nm, 0.050 max. (280nm, 0.1M H2O), 0.050 max. (260nm
Melting Point 206°C to 211°C
Color White
pH 10.9 to 11.6 (1% aq. soln. at 25°C)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Solubility Information (10% in water) Clear colorless
Formula Weight 259.3
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material CAPSO sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • CAPSO sodium salt
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.