missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals CAPSO sodium salt, 98%

Product Code. 11480678
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480678 25 g 25g
11490678 100 g 100g
11400688 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480678 Supplier Thermo Scientific Chemicals Supplier No. B25202.14
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 102601-34-3 | C9H12NO4S | 230.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102601-34-3
Molecular Formula C9H12NO4S
Molecular Weight (g/mol) 230.26
MDL Number MFCD00070063
InChI Key CLEVULOKVPBHCU-VIFPVBQESA-M
Synonym capso sodium salt, sodium 3-cyclohexylamino-2-hydroxypropane-1-sulfonate, 3-cyclohexylamino-2-hydroxypropanesulfonic acid sodium salt, capso sodium, capso, 0.2m buffer solution, ph 8.5, capso, 0.2m buffer solution, ph 9.0, capso, 0.2m buffer solution, ph 9.5, sodium 3-cyclohexylamino-2-hydroxypropanesulfonate, sodium 3-cyclohexylamino-2-hydroxy-propane-1-sulfonate, potassium 3-cyclohexylamino-2-hydroxypropane-1-sulfonate
PubChem CID 25000353
IUPAC Name sodium;3-(cyclohexylamino)-2-hydroxypropane-1-sulfonate
SMILES O[C@@H](CNC1=CC=CC=C1)CS([O-])(=O)=O

Specifications

Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 259.3
Percent Purity 98%
Chemical Name or Material CAPSO sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.