Learn More
Erlab™ Captair by Erlab™ Benchcaps
Product Code. 15395102 Shop All Erlab Products
Change view
Click to view available options
For Use With (Equipment):
CaptairFlex XLS633 Ductless Fume Hood
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15395102 CaptairFlex XLS633 Ductless Fume Hood Each
1 options
Product Code. 15395102 Supplier Erlab™ Supplier No. I0519000302
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Use these Erlab™ benchcaps with CaptairFlex ductless fume hoods.
  • Fixed metal stand for CaptairFlex™ ductless fume hoods
  • Equipped with 4 cylinders to upgrade the unit
Specifications
Product Type Benchcap
Product Line Captair
For Use With (Equipment) CaptairFlex XLS633 Ductless Fume Hood