Learn More
Erlab™ Captair by Erlab™ Stainless Steel Work Surfaces
Produktkod. 13416583
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
CaptairFlow 391 Ductless Fume Hood
CaptairFlow 483 Ductless Fume Hood
CaptairFlow 714 Ductless Fume Hood
Förpackningsstorlek
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. For Use With (Equipment) unitSize
13416583 CaptairFlow 391 Ductless Fume Hood Each
13436583 CaptairFlow 483 Ductless Fume Hood Each
13426583 CaptairFlow 714 Ductless Fume Hood Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 13416583 missing translation for 'mfr' Erlab™ missing translation for 'supplierNo' I0116050001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Denna artikel kan inte returneras. Se returpolicy
Designed for laboratories specializing in fields such as biology, botany, aerospace, electronics, pharmaceutical and cosmetics. Erlab™ Captair™ Stainless Steel Work Surfaces are for use with Captair PCR work stations.
  • High chemical and mechanical resistance
  • Rounded corners to facilitate cleaning operations
  • Built in spill tray
Specifikationer
Material Stainless Steel
For Use With (Equipment) CaptairFlow 391 Ductless Fume Hood
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.