Learn More
Erlab™ Captair by Erlab™ V201 Version 04 Filtration Columns
Product Code. 12187730
Change view
Click to view available options
Filter Type:
AS Carbon Filter
BE Carbon Filter
F Carbon Filter
K Carbon Filter
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Filter Type unitSize
12187730 BE Carbon Filter Each
12147730 AS Carbon Filter Each
12038558 F Carbon Filter Each
12018568 K Carbon Filter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12187730 Supplier Erlab™ Supplier No. G0204/BE
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Product Type Filtration Column
Filter Type BE Carbon Filter
For Use With (Application) For storage with Organic Vapors and Acids