Learn More
Erlab™ Captair Smart Trespa™ Work Surface
Erlab™ Captair Smart Trespa™ Work Surface is designed to be used with the range of Captair Smart Ductless Filtering Fume Hoods.
964.60 € - 1770.20 €
Specifications
Includes | Spill Tray |
---|---|
Product Type | Work Surface |
Material | Phenolic Resin |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15545154
|
Erlab™
I0160030001 |
Smart 321 Fume Hood |
964.60 €
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15575154
|
Erlab™
I0161030001 |
Smart 391 Fume Hood |
1028.20 €
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15515164
|
Erlab™
I0163030001 |
Smart 392 Fume Hood |
1150.10 €
Each |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15505164
|
Erlab™
I0162030001 |
Smart 481 Fume Hood |
1187.20 €
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15535164
|
Erlab™
I0164030001 |
Smart 483 Fume Hood |
1325.00 €
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15565164
|
Erlab™
I0165030001 |
Smart 633 Fume Hood |
1505.20 €
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15585164
|
Erlab™
I0166030001 |
Smart 714 Fume Hood |
1770.20 €
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Made of phenolic resin with ergonomic arm for a comfortable working position
- High chemical and mechanical resistance
- Ideal for precision weighing operations
Specifications
Spill Tray | |
Work Surface | |
Phenolic Resin |