Learn More
Erlab™ Captair™ Store™ Filter Alarm
Brand:  Erlab™ I0299000001
Code : 50
Additional Details : Weight : 0.80000kg
Product Code. 12028992
  • 1192.50 € / Each
Estimated Shipment: 23-09-2024
to see stock.
Specifications
For solvents (M and XLS fume hoods and Captair™ Store™)
Filter Saturation Electronic Alarm Molecode S
For Auto Detection of Filter Saturation by Solvents