missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Capto™ adhere ImpRes Multimodal BioProcess™ Chromatography Medium

Product Code. 15111556 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15111556 100 mL 100mL
15121556 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15111556 Supplier Cytiva Supplier No. 17371502

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Includes

Certificate of Analysis

For high-resolution polishing of monoclonal antibodies (MAbs) and other biomolecules

The strong anion exchange multimodal ligand displays high selectivity compared with traditional ion exchanger, which allows the possibility to solve challenging purification problems. Main contaminants in MAb processes such as DNA, host cell proteins (HCP), leached protein A, aggregates, and viruses are efficiently separated.

  • High-resolution beads and ligand selectivity ensures high yield
  • Efficient removal of aggregates, viruses, and main contaminants in MAb processes
  • Enables use of platform approach to MAb process development
  • Allows separation of MAb charged variants

Avoid using oxidizing agents, anionic detergents.

TRUSTED_SUSTAINABILITY

Especificaciones

Quantity 100 mL
Stability 1 M Acetic Acid, 1 M NaOH, 2 M NaCl, 5% 1-Propanol, 30% Isopropanol, 70% Ethanol
For Use With (Application) For High Resolution Polishing of Monoclonal Antibodies (MAbs) and Other Biomolecules
Storage Requirements 4°C to 30°C
Anion Exchange Capacity 0.08 to 0.11 mM Cl-/mL
Binding Capacity 45 to 85 mg Mab/mL
Product Type Capto Adhere ImpRes Media
Particle Size 40 μm
Includes Certificate of Analysis
Max. Pressure 300 kPa
Matrix High-flow Agarose
pH Range 3 to 12
Storage Buffer 20% Ethanol
Ligand Type Multimodal Strong AnIon Exchanger
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • Capto adhere ImpRes, 100 ml
Palabra de advertencia
  • Warning
Clasificación
  • Flammable liquid Category 3
Indicaciones de peligro
  • H226-Flammable liquid and vapour.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Otros peligros
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.