missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Capto™ Adhere Media

Product Code. 10792852 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10792852 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10792852 Supplier Cytiva Supplier No. 17544410
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A multimodal anion exchanger to follow the Protein A capture step, making for a streamlined process

This product is designed to allow for a two-step process with purification of monoclonal antibodies and Protein A capture, which cuts cost and time.

  • Two-step chromatographic process with protein A affinity
  • Multimodal anion exchanger for post-protein A purification of monoclonal antibodies
  • Wide operational window of pH and conductivity
  • High capacity and productivity in flow-through mode
  • Average particle size of 75μm
  • Highy crossed-linked agarose
  • Ligand N-benzyl-n-methyl ethanolamine

For intermediate purification and polishing of monoclonal antibodies after the Protein A capture step.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mL
Stability 1 M Acetic Acid, 1 M Sodium Hydroxide
For Use With (Application) For Multimodal Chromatography
Storage Requirements 4°C to 30°C
Product Type Capto Adhere Media
Flow Rate 600 cm/Hoμr
Particle Size 75 μm
Height (English) Bed 7.87 in.
Height (Metric) Bed 20 cm
Ionic Capacity 0.09 to 0.12 mM Cl-/mL
Includes Certificate of Analysis
Ion Exchange Type Multimodal Strong AnIon Exchanger
Max. Pressure 300 kPa
Matrix Highly Cross-Linked Agarose
pH Range 2 to 14
Storage Buffer 20% Ethanol
Ligand Type N-benzyl-n-methyl ethanolamine
Show More Show Less
compliance-icons
Product Identifier
  • Capto adhere
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.