missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Capto™ Core 700
Shop All Cytiva Products
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
Each
Description
The novel core bead technology and multimodal, octylamine ligand give Capto Core 700 dual functionality, size exclusion, and binding chromatography in one chromatography medium (resin).
- For intermediate purification and polishing of viruses and other large biomolecules (Mr > 700 000) in flowthrough mode.
- Novel core bead technology allows efficient capture of contaminants while target molecules are collected in the flowthrough.
- Significantly improved productivity and higher flow rates compared with gel filtration methods.
- Straightforward optimization due to flowthrough chromatography and robust performance.
- Convenient small-scale purification, process development, and scale-up using prepacked HiTrap and HiScreen columns.
- Capto Core 700 is composed of a ligand-activated core and inactive shell.
- The inactive shell excludes large molecules (cut off ∼ Mr 700000 [700 kDa]) from entering the core through the pores of the shell.
- These larger molecules are collected in the column flowthrough while smaller impurities bind to the internalized ligands.
- The core of each bead is functionalized with ligands that are both hydrophobic and positively charged, resulting in a highly efficient multimodal binding of various contaminants small enough to enter the core.
- The multimodal ligands ensure strong binding with most impurities over a wide range of pH and salt concentrations.
- Bound impurities are removed from the beads by cleaning-in-place (CIP) procedures using NaOH and in most cases, a solvent.
Specifications
Specifications
| Buffer | 1M NaOH, 6M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol |
| Product Type | Capto Core 700 chromato graphy media |
| Quantity | 100 mL |
| Flow Rate | <500 cm/hr. |
| Ligand Type | Octylamine |
| pH Range | 2 to 14 |
| Particle Size | 90μm |
| Support Type | Highly Cross-linked Agarose |
| Content And Storage | 4° to 30°C |
| For Use With (Application) | Intermediate purification and polishing of viruses and other large biomolecules |
| Show More |
Safety and Handling
Product Identifier
- Capto Core 700, 100 ml
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction