missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Capto™ HiRes Ion Exchange Chromatography Columns

Product Code. 16287791 Shop All Cytiva Products
Change view
Click to view available options
Type:
Capto™ HiRes Q 10/100 Column
Capto™ HiRes Q 5/50 Column
Capto™ HiRes S 10/100 Column
Capto™ HiRes S 5/50 Column
Column Capacity:
1 mL
8 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Column Capacity Unit Size
16287791 Capto™ HiRes Q 10/100 Column 8 mL Each
16267791 Capto™ HiRes Q 5/50 Column 1 mL Each
16257791 Capto™ HiRes S 5/50 Column 1 mL Each
16277791 Capto™ HiRes S 10/100 Column 8 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16287791 Supplier Cytiva Supplier No. 29275881
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prepacked columns for high-resolution purification of proteins and other biomolecules by ion exchange chromatography. They are well-suited for the preparation of highly pure proteins for structural studies and protein analysis.

  • High resolution columns for laboratory scale ion exchange chromatography
  • Suitable for preparing highly pure proteins, and protein analysis in research or quality control applications
  • High quality column with excellent reproducibility, durability, and high chemical and pH stabilities
  • Replacement for Mono Q™ and Mono S™ columns delivering similar or improved resolution, with easy transition from one column platform to the other
  • Chromatography packing resins are based on a high-flow agarose base matrix and a small bead size (9 μm average)
TRUSTED_SUSTAINABILITY

Specifications

Bed Height 100 mm
Column Type Ion Exchange Column
Description Ionic Capacity: ∼0.23 mmol Cl-/mL resin
Column Capacity 8 mL
Flow Rate 0.5 to 2 mL/min.
For Use With (Application) Anaylsis or Purification of Proteins and Nucleic Acids
Ion Exchange Group Quaternary Ammonium
Max. Pressure 4 MPa (40 bar)
Particle Size ∼9μm
Product Line Capto
Type Capto™ HiRes Q 10/100 Column
Material Tricorn™ Glass
Packing Material Highly Cross-Linked Agarose
Particle Shape Spherical
pH 2 to 12
Quantity 1 Ea.
Temperature (Metric) 4°C to 40°C
Diameter (Metric) Inner 10 mm
Show More Show Less
compliance-icons
Product Identifier
  • Capto HiRes Q 10/100
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.