missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Capto™ MMC ImpRes Multimodal BioProcess™ Chromatography Medium

Product Code. 15141556 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15141556 100 mL 100mL
15131556 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15141556 Supplier Cytiva Supplier No. 17371602
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For high-resolution polishing of monoclonal antibodies (MAbs) and other biomolecules

  • High yields achieved through high-resolution beads and ligand selectivity
  • Efficient removal of aggregates, viruses, and main contaminants in MAb processes
  • Enables use of platform approach to MAb process development
  • Displays broad pH/salt operational window

Avoid using oxidizing agents, cationic detergents.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mL
Stability 1 M Acetic Acid, 1 M NaOH, 2 M NaCl, 5% 1-Propanol, 30% Isopropanol, 70% Ethanol
For Use With (Application) For High Resolution Polishing of Monoclonal Antibodies (MAbs) and Other Biomolecules
Storage Requirements 4°C to 30°C
Anion Exchange Capacity 25 to 39 μM/mL
Binding Capacity 60 to 90 mg Mab/mL
Product Type Capto MMC ImpRes Media
Particle Size 40 μm
Max. Pressure 300 kPa
Matrix High-flow Agarose
pH Range 3 to 12
Storage Buffer 20% Ethanol, 0.2 M Sodium Acetate
Ligand Type Multimodal Weak CatIon Exchanger
Format Bulk Media
Show More Show Less
compliance-icons
Product Identifier
  • Capto MMC ImpRes
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.