missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Capto S, 25mL

Product Code. 10829450 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10829450 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10829450 Supplier Cytiva Supplier No. 17544110
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed to meet the demands of modern large-scale manufacturers. Cytiva Capto S, 25ml is a strong cation exchange BioProcess medium for fast, efficient, and cost-effective capture and intermediate protein purification of sulfonate group ligands.

Capto S is designed for capture and intermediate purification of proteins from large feed volumes. It is composed of a rigid, high-flow agarose matrix modified with dextran surface extenders and a strong sulfonate (S) cation exchanger. The combination of high volume throughput and high dynamic binding capacity for Capto S raises the productivity in industrial downstream processes. Capto S withstands effective and rigorous CIP procedure and the chemical stability ensures long media lifetime.
  • Strong cation exchanger for high-productivity capture and intermediate purification when high volume throughput is essential
  • High dynamic binding capacity at high flow raises productivity
  • High-volume throughput cuts process times
  • Cost-effective processing with smaller unit operations
  • Meets industrial demands: security of supply, technical and regulatory support
TRUSTED_SUSTAINABILITY

Specifications

Description Capto S, 25 mL
Matrix Highly cross-linked agarose with dextran surface extender
For Use With (Application) Capto S is a strong cation exchanger for fast, efficient, and cost-effective capture and intermediate protein purification from large feed volumes.
Ligand Type Sulfonate group
Content And Storage 4°C to 30°C, 20% Ethanol, 0.2 M Sodium Acetate
Particle Size 90 μm, d50v is the median particle size of the cumulative volume distribution.
pH 4 to 12
Quantity 25 mL
compliance-icons
Product Identifier
  • capto S
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.