missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Capto S™ ImpAct Chromatography Media
Shop All Cytiva Products
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
Description
- High binding capacity, > 100 mg MAb/mL medium.
- Efficient aggregate removal at high load of monoclonal antibodies.
- High-resolution polishing based on the well-established Capto platform.
- Flexibility of design thanks to a large operational window of flow rates and bed heights for easy optimization and scaling.
- High productivity enabling cost-efficient manufacturing.
- Security of supply and comprehensive regulatory support.
Specifications
Specifications
| Product Type | Capto S ImpAct Chromatography Media |
| Quantity | 25 mL |
| Material | High-flow agarose |
| pH Range | 4 to 12 |
| Particle Size | 50μm |
| Matrix | Ion Exchange Chromatography Media |
Safety and Handling
Product Identifier
- Capto S ImpAct, 25 ml
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Ethanol
- UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction