missing translation for 'onlineSavingsMsg'
Learn More

Captopril, MedChemExpress

Product Code. 16472116
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472116 500 mg 500mg
16408266 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16472116 Supplier Medchem Express Supplier No. HYB0368
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Captopril (SQ 14225), antihypertensive agent, is a thiol-containing competitive, orally active angiotensin-converting enzyme (ACE) inhibitor (IC50=0.025 μM) and has been widely used for research of hypertension and congestive heart failure. Captopril is also a New Delhi metallo-β-lactamase-1 (NDM-1) inhibitor with an IC50 of 7.9 μM.

Captopril (SQ 14225), antihypertensive agent, is a thiol-containing competitive, orally active angiotensin-converting enzyme (ACE) inhibitor (IC50=0.025 μM) and has been widely used for research of hypertension and congestive heart failure. Captopril is also a New Delhi metallo-β-lactamase-1 (NDM-1) inhibitor with an IC50 of 7.9 μM.

Specifications

Chemical Name or Material Captopril
CAS 62571-86-2
Color White
Purity Grade Notes Research
Molecular Formula C9H15NO3S
Quantity 500 mg
Synonym SQ 14225
Solubility Information DMSO : ≥ 50 mg/mL (230.11 mM) ∣H2O : ≥ 50 mg/mL (230.11 mM)
SMILES O=C(O)[C@H]1N(C([C@H](C)CS)=O)CCC1
Molecular Weight (g/mol) 217.29
Formula Weight 217.29
Percent Purity 99.2%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.