Learn More
Captopril
Description
Captopril has also been shown to inhibit the formation of angiotensin II, a bioactive peptide that stimulates angiogenesis and increases microvessel density. Captopril demonstrates noncompetitive inhibition of tyrosinase monophenolase activity and competitive inhibition of diphenolase activity
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Captopril has also been shown to inhibit the formation of angiotensin II, a bioactive peptide that stimulates angiogenesis and increases microvessel density. Captopril demonstrates noncompetitive inhibition of tyrosinase monophenolase activity and competitive inhibition of diphenolase activity
Solubility
Soluble in water (160 mg/ml) at 25°C, sesame and corn oils (<1mg/mL) at 25°C, methanol (>100 mg/ml), ethanol (>100 mg/ml), isopropanol (>100 mg/ml), chloroform (>100 mg/ml), and dichloromethane (>100 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 104°C to 108°C |
| Color | White |
| Quantity | 25 g |
| Beilstein | 477887 |
| Merck Index | 14,1774 |
| Solubility Information | Soluble in water (160mg/ml) at 25°C,sesame and corn oils (<1mg/ml) at 25°C,methanol (>100mg/ml),ethanol (>100mg/ml),isopropanol (>100mg/ml),chloroform (>100mg/ml),and dichloromethane (>100mg/ml). |
| Formula Weight | 217.29 |
| Physical Form | Powder |
| Chemical Name or Material | Captopril |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.