missing translation for 'onlineSavingsMsg'
Learn More

Captopril, TRC

Product Code. 30138124
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138124 100 g 100g
17949619 10 g 10g
18063710 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138124 Supplier Toronto Research Chemicals Supplier No. TRCC175750100G
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62571-86-2
Molecular Formula C9 H15 N O3 S
Molecular Weight (g/mol) 217.29
Synonym L-Proline, 1-(3-mercapto-2-methyl-1-oxopropyl)-, (S)-, 1-[(2S)-3-Mercapto-2-methyl-1-oxopropyl]-L-proline, (-)-1-[(2S)-3-Mercapto-2-methyl-1-oxopropyl]-L-proline, (-)-Captopril, Acediur, Aceplus, Acepress, Acepril, Aceten, Aceten 25, Alopresin, Angiopril, Capoten, Capotril, CaptoHexal, Captolane, Captomax, Capton, Captopril, Captopril GP, Captopril Generis, Captopril Mepha, Captopril ratiopharm, Captoril, Captotec, Cesplon, Dilabar, Facopril, Farcopril, Garranil, Hipertil, Hypopress, L-Captopril, Lopirin, Lopril, Lotensine, Novocaptopril, S-Captopril, SA 333, SQ 14225, Tensiomin, Tensobon, Tensoprel, Zapto
IUPAC Name (2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylic acid
SMILES C[C@H](CS)C(=O)N1CCC[C@H]1C(=O)O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C9H15NO3S/c1-6(5-14)8(11)10-4-2-3-7(10)9(12)13/h6-7,14H,2-5H2,1H3,(H,12,13)/t6-,7+/m1/s1
Formula Weight 217.0773
Chemical Name or Material Captopril
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.