Learn More
Thermo Scientific™ CaptureSelect™ AAT-XL RoboColumn™ Pre-packed columns
Shop All Thermo Scientific ProductsDescription
CaptureSelect RoboColumn™ are small chromatography columns used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.
The CaptureSelect AAT-XL Affinity Matrix in this RoboColumn has been specifically designed for the purification of native human alpha-1 antitrypsin (AAT) from complex sources like plasma/serum and recombinant human AAT from cell culture supernatants. The high selectivity and yield obtained using CaptureSelect AAT-XL Affinity Matrix enable a robust and efficient purification process with excellent purity obtained in a single step.
Features of this affinity matrix include:
- Highly selective for native and recombinant human AAT
- High recovery and yield obtained in a single purification step
- Mild elution at pH 6
- Excellent scalability
- Non-animal-derived
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Characteristics
- Column size: 200 μL, 1 strip, 8 columns (8-row)
- Column dimension: 5-mm diameter, 10-mm bed height
- Matrix: agarose-based, epoxide-activated
- Average particle size: 65 μm
- Ligand: CaptureSelect AAT-XL affinity ligand
- Ligand coupling: epoxide coupling
- Binding capacity: 15 mg AAT/mL resin depending on flow rate, column height, and contact time
- Elution conditions: 20 mM Tris, 2.0 M MgCl2, pH 6.0
- Formulation buffer: 20% (v/v) ethanol
- Operating pressure: 2 bar (maximum)
- Flow rate: 16–150 cm/hr
RoboColumn is a trademark of Repligen.
Specifications
Specifications
| Max. Pressure | 2 bar |
| Pore Size | Not Applicable |
| Column Capacity | 200 μL |
| Length (Metric) | 1 cm |
| Quantity | 1 Strip, 8 Columns |
| Particle Size | 65 μm |
| For Use With (Equipment) | Chromatography Systems |
| Type | Affinity Column |
| Diameter (Metric) Inner | 0.5 cm |
| Content And Storage | Store at 2°C to 8°C. Protect from light. Do not freeze. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.