missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Antithrombin III Affinity Matrix

Product Code. p-7180803 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
Unit Size:
10mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18456548 5 mL 5mL
18456588 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18456548 Supplier Thermo Scientific™ Supplier No. 190317005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This is one of a group of Thermo Scientific™ CaptureSelect™ protein purification affinity resins developed for the simple, single-step purification of a variety human plasma and serum proteins.

This is one of a group of CaptureSelect protein purification affinity resins developed for the simple, single-step purification of a variety human plasma and serum proteins. This unique selection of affinity purification resins can purify proteins or other non-antibody biomolecules from either recombinant or native sources with a high degree of affinity and specificity, and are immobilized on a high capacity agarose support.

Specifications

Content And Storage Store at 2°C to 8°C.
Form Liquid Suspension
Product Line CaptureSelect
Quantity 5 mL
Type Bulk Resin
Stationary Phase Anti Antithrombin III
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.