missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-AAVX Conjugate

Product Code. 18486448 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
500 μg
Unit Size:
100µg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18486448 100 μg 100µg
18446738 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18486448 Supplier Thermo Scientific™ Supplier No. 7103522100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ Biotin Anti-AAVX Conjugate consists of a 14 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to native and recombinant adeno-associated virus (AAV) particles of various serotypes.

CaptureSelect Biotin Anti-AAVX Conjugate consists of a 14 kDa recombinant single domain antibody fragment that specifically binds with high affinity and selectivity to native and recombinant adeno-associated virus (AAV) particles of various serotypes.

The anti-AAVX affinity ligand has demonstrated binding reactivity towards a large set of different AAV serotypes including AAV1 to AAV8, and AAVrh10, indicating that an even broader range of AAV serotypes could potentially be recognized and detected. The affinity ligand is chemically conjugated to biotin via a spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

This conjugate enables the detection, quantitation, and characterization of native and recombinant AAV virus particles. Applications for CaptureSelect Biotin Anti-AAVX Conjugate include ELISA, immunoprecipitation with streptavidin-coupled Dynabeads, Gyrolab™-based immunoassays, and use with label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -5°C to -30°C.
Shipping Condition Dry Ice
Form Liquid
Product Line CaptureSelect
Quantity 100 μg
Type Conjugate
Ligand Type Antibody
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.