missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-FIX Conjugate

Product Code. 18476778 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
500 μg
Unit Size:
100µg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18476778 100 μg 100µg
18426538 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18476778 Supplier Thermo Scientific™ Supplier No. 7103002100
  • ONLINE EXCLUSIVE PRICE
    575.00 €
    540.00 € / 100µg
    Save 35.00 €
    6% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ Biotin Anti-FIX Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for both recombinant and plasma derived human Factor IX (FIX).

CaptureSelect Biotin Anti-FIX Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for both recombinant and plasma derived human Factor IX (FIX). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect Biotin Anti-FIX Conjugate allows you to:

  • Detect, quantitate, and characterize recombinant and plasma derived human FIX

Applications for CaptureSelect Biotin Anti-FIX Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

TRUSTED_SUSTAINABILITY

Specifications

Product Type Biotin Anti-FIX Conjugate
Conjugate Biotin and Other Haptens
Quantity 100 μg
Product Line CaptureSelect
Content And Storage Store in freezer (-5°C to -30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.