missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-LC-lambda (Human) Conjugate

Product Code. 18446538 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 μg
500 μg
Unit Size:
100µg
500µg
This item is not returnable. View return policy

Product Code. 18446538

Brand: Thermo Scientific™ 7103082100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ Biotin Anti-LC-lambda (Hu) Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the constant domain of human lambda light chains.

CaptureSelect Biotin Anti-LC-lambda (Hu) Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the constant domain of human lambda light chains. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect Biotin Anti-LC-lambda (Hu) Conjugate allows you to:

  • Detect, quantitate, and characterize all human antibody isotypes and subclasses (for example, IgG1 to 4, IgM, IgA, IgD, and IgE), and Fab fragments thereof, that contain a lambda light chain; as well as recombinant human Fab fragments and/or Fab and Fab2 fragments obtained after papain or pepsin digestion
  • Avoid cross-binding with human antibodies that possess a kappa light chain or IgG from, for example, mouse, rat, or rabbit
  • Screen antibody-antigen interactions—biotin anti-LC-lambda (Hu) forms a stable [low k(diss)] complex with, for example, human IgG antibodies through bivalent binding

Applications for CaptureSelect Biotin Anti-LC-lambda (Hu) Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Content And Storage Store at -5°C to -30°C.
Form Liquid
Product Line CaptureSelect
Quantity 100 μg
Target Lambda Light Chain
Type Conjugate
Label or Dye Biotin & Other Haptens
Ligand Type Antibody
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.