missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ CD81 Magnetic Agarose Beads

Product Code. 30276103 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 x 1 mL
1 x 5 mL
1 x 50 mL
Unit Size:
1mL
50mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30276103 1 x 50 mL 50mL
30275819 1 x 1 mL 1mL
30275477 1 x 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30276103 Supplier Thermo Scientific™ Supplier No. 2883842050
  • ONLINE EXCLUSIVE PRICE
    8610.00 €
    8490.00 € / 50mL
    Save 120.00 €
    1% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CaptureSelect CD81 Magnetic Agarose Beads are high-capacity and high-throughput magnetic affinity particles for exosome purification using manual or robotic magnetic separators.

CaptureSelect CD81 Magnetic Agarose Beads are high-capacity and high-throughput magnetic affinity particles for exosome purification using manual or robotic magnetic separators. The CaptureSelect CD81 affinity ligand recognizes the tetraspanin protein CD81 on the surface of human extracellular vesicles (EVs) like exosomes, enabling purification of CD81-expressing exosomes. CD81-expressing exosomes can be directly isolated from complex source materials in a single step with high purity and yield.

Features of CaptureSelect CD81 Magnetic Agarose Beads include:

  • One-step human CD81-expressing exosome purification with high purity from crude cell clarified supernatant, TFF-concentrated cell lysates, and blood plasma
  • Compatible with manual and automated workflows, e.g., KingFisher instruments
  • Short operation time
  • No sample dilution required during isolation; repetitive sample loading feasible
  • Relatively mild elution at pH 9, resulting in efficient release of exosomes from the magnetic beads.
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in yeast Saccharomyces cerevisiae by a production process free of any animal components (animal origin-free).

Main characteristics

Matrix: 6% cross-linked magnetized agarose

Average particle size: 25-30 μm

Suspension: 25% slurry

Ligand: CaptureSelect CD81 affinity ligand

Binding capacity: estimated at ≥ 1 x 1010 EVs /mL of magnetic bead slurry

Elution conditions: 50 mM Tris, 1 M arginine, pH 9

Formulation buffer: Tris-buffered saline, 0.05% Tween 20, 0.05 % sodium azide

Specifications

Formulation 25% (v/v) slurry in Tris buffered saline with 0.05% Tween 20 and 0.05% sodium azide
Matrix 6% cross-linked magnetized agarose
Content And Storage Store at 2°C to 8°C
Shipping Condition Wet Ice
Particle Size 25 to 30 μm
Form Resin Slurry
Product Line CaptureSelect
Quantity 1 x 50 mL
Type Magnetic Agarose Bead
Ligand Type CD81 affinity ligand
Sufficient For Sample Preparation
Binding Capacity ∼1x1010 EVs/mL of Magnetic Beads
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.