missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ CH1-XL GoPure™ Column

Product Code. 30275601 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Column
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30275601 1 Column 5mL
30276303 1 Column 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30275601 Supplier Thermo Scientific™ Supplier No. 6943462005
  • ONLINE EXCLUSIVE PRICE
    1255.00 €
    1200.00 € / 5mL
    Save 55.00 €
    4% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CaptureSelect CH1-XL GoPure Columns are ready-to-use columns designed for fast and easy purification of human IgG on standard chromatography systems with high purity and yield in a single step.

CaptureSelect CH1-XL GoPure Columns are ready-to-use columns designed for fast and easy purification of human IgG on standard chromatography systems with high purity and yield in a single step.

The pre-packed column contains CaptureSelect CH1-XL Affinity Matrix, which recognizes the CH1 domain of human IgG antibodies, enabling purification of all subclasses (1, 2, 3, and 4) of human IgG and Fab fragments, independent of the light-chain isotype and source material.

Features of this affinity matrix include:

  • Platform approach for purification of IgGs and their Fab fragments directly from complex source materials in a single step with high purity and yield
  • Elution possible at pH 4.0–4.5 with 50 mM acetate buffer
  • No co-purification of overproduced free light chains or light chain dimers
  • Specific for human antibodies; works in the presence of serum (bovine/horse)
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created using a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

Matrix: agarose-based, epoxide-activated

Average particle size: 65 μm

Ligand: CaptureSelect CH1-XL affinity ligand

Ligand coupling method: epoxide coupling

Binding capacity: >15 mg/mL Fab's depending on flow rate, column height, contact time

Elution conditions: 50 mM sodium acetate buffer, pH 4.0–4.5

Column dimensions: 1 mL column: 7 x 25 mm; 5 mL column: 16 x 25 mm

Max. recommended flow: 1 mL column: 6 mL/min; 5 mL column: 10 mL/min

Max. operation pressure: 2 bar

Formulation buffer: 20% (v/v) ethanol

Specifications

Max. Pressure 0.3 Mpa
Pore Size Not Applicable
Column Capacity 5 mL
Length (Metric) 2.5 cm
Quantity 1 Column
Particle Size 65 μm
For Use With (Equipment) Chromatography Systems
Type Affinity Column
Diameter (Metric) Inner 1.6 cm
Content And Storage Store at 2°C to 8°C. Protect from light. Do not freeze.
Binding Capacity >15 mg/mL human Fab fragment
Product Line CaptureSelect
Matrix Agarose
Shipping Condition Wet Ice
Column Format Pre-packed
Bed Volume 5 mL
Bed Height 25 mm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.