missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ CH1-XL Ligand Leakage ELISA

Product Code. 18466748 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Assay
10 Assays
Unit Size:
Each
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18466748 1 Assay Each
18426558 10 Assays Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18466748 Supplier Thermo Scientific™ Supplier No. 810346201
  • ONLINE EXCLUSIVE PRICE
    1110.00 €
    1068.00 € / Pack of 1
    Save 42.00 €
    3% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ CH1-XL Ligand Leakage ELISA Kit is designed to detect possible leached ligand of antibodies or antibody fragments purified using CaptureSelect CH1-XL Affinity Matrix.

CaptureSelect CH1-XL Ligand Leakage ELISA Kit is designed to detect possible leached ligand of antibodies or antibody fragments purified using CaptureSelect CH1-XL Affinity Matrix. The ligand, a 14-kD recombinant protein, is covalently coupled for high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect CH1-XL Ligand Leakage ELISA Kit can be used as a tool to aid in development of an optimal purification process and in routine quality control of in-process streams and final product.

Specifications

Content And Storage Store at -20°C.
Detection Method Absorbance at 450 nm
Immunoassay Kit Format Sandwich ELISA Kit
Product Line CaptureSelect
Product Type Ligand Leakage ELISA
Sample Type Ligand Leachate
Target Species Not Applicable
Quantity 1 Assay
Shipping Condition Dry Ice
Target CaptureSelect CH1-XL Affinity Ligand
Detection Limit 1 ng/mL
Format Individual components
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.